missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(Hydroxymethyl)furfural, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 121460010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-(Hydroxymethyl)furfural | |
30.0°C to 34.0°C | |
114°C to 116°C (1.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5627 | |
18, V,1, 130 | |
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole | |
NOEGNKMFWQHSLB-UHFFFAOYSA-N | |
5-(hydroxymethyl)furan-2-carbaldehyde | |
237332 | |
126.11 | |
Crystalline Powder, Chunks or Liquid |
67-47-0 | |
Yellow | |
79°C | |
97.5% min. (GC) | |
C6H6O3 | |
1 g | |
15,4871 | |
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents | |
C1=C(OC(=C1)C=O)CO | |
126.11 | |
CHEBI:412516 | |
98% |
Chemical Identifiers
67-47-0 | |
126.11 | |
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole | |
CHEBI:412516 | |
C1=C(OC(=C1)C=O)CO |
C6H6O3 | |
NOEGNKMFWQHSLB-UHFFFAOYSA-N | |
237332 | |
5-(hydroxymethyl)furan-2-carbaldehyde |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 200-654-9
RUO – Research Use Only