missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)furfural, 98%

CAS: 67-47-0 | C6H6O3 | 126.11 g/mol

$109.36 - $1337.37

Chemical Identifiers

CAS 67-47-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
InChI Key NOEGNKMFWQHSLB-UHFFFAOYSA-N
Synonym 5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
PubChem CID 237332
ChEBI CHEBI:412516
IUPAC Name 5-(hydroxymethyl)furan-2-carbaldehyde
SMILES C1=C(OC(=C1)C=O)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC121460010
SDS View Documents
Thermo Scientific Chemicals
121460010
1 g Glass bottle
Each for $109.36
Only null left
 
AC121460050
SDS View Documents
Thermo Scientific Chemicals
121460050
5 g Glass bottle
Each for $314.09
Only null left
 
AC121460250
SDS View Documents
Thermo Scientific Chemicals
121460250
25 g Glass bottle
Each for $1,337.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-47-0
126.11
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
CHEBI:412516
C1=C(OC(=C1)C=O)CO
C6H6O3
NOEGNKMFWQHSLB-UHFFFAOYSA-N
237332
5-(hydroxymethyl)furan-2-carbaldehyde

Specifications

67-47-0
Yellow
79°C
97.5% min. (GC)
C6H6O3
1 g
15,4871
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
C1=C(OC(=C1)C=O)CO
126.11
CHEBI:412516
98%
5-(Hydroxymethyl)furfural
30.0°C to 34.0°C
114°C to 116°C (1.0 mmHg)
Authentic
Glass bottle
1.5627
18, V,1, 130
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
NOEGNKMFWQHSLB-UHFFFAOYSA-N
5-(hydroxymethyl)furan-2-carbaldehyde
237332
126.11
Crystalline Powder, Chunks or Liquid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 200-654-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only