missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Hydroxymethyl)furfural, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 121460050

 View more versions of this product

Catalog No. AC121460050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-(Hydroxymethyl)furfural
30.0°C to 34.0°C
114°C to 116°C (1.0 mmHg)
Authentic
Glass bottle
1.5627
18, V,1, 130
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
NOEGNKMFWQHSLB-UHFFFAOYSA-N
5-(hydroxymethyl)furan-2-carbaldehyde
237332
126.11
Crystalline Powder, Chunks or Liquid
67-47-0
Yellow
79°C
97.5% min. (GC)
C6H6O3
5 g
15,4871
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
C1=C(OC(=C1)C=O)CO
126.11
CHEBI:412516
98%

Chemical Identifiers

67-47-0
126.11
5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
CHEBI:412516
C1=C(OC(=C1)C=O)CO
C6H6O3
NOEGNKMFWQHSLB-UHFFFAOYSA-N
237332
5-(hydroxymethyl)furan-2-carbaldehyde
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 200-654-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only