missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC26701DA

 View more versions of this product

Catalog No. CC26701DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Methyl-2-phenyl-1, 3-oxazole-4-carboxylic acid
97%
C11H8NO3
1g
YABCPNYCFFUVNM-UHFFFAOYSA-M
5-methyl-2-phenyl-1,3-oxazole-4-carboxylic acid
318533
97%
18735-74-5
Amber Glass Bottle
MFCD00275475
5-methyl-2-phenyloxazole-4-carboxylic acid, 2-phenyl-5-methyloxazole-4-carboxylic acid, 5-methyl-2-phenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazole-4-carboxylicacid, 4-carboxy-5-methyl-2-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid,5-methyl-2-phenyl, 2-phenyl-5-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylicacid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazol-4-ylcarboxylic acid
CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O
202.19
203.2

Chemical Identifiers

18735-74-5
202.19
YABCPNYCFFUVNM-UHFFFAOYSA-M
318533
CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O
C11H8NO3
MFCD00275475
5-methyl-2-phenyloxazole-4-carboxylic acid, 2-phenyl-5-methyloxazole-4-carboxylic acid, 5-methyl-2-phenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazole-4-carboxylicacid, 4-carboxy-5-methyl-2-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid,5-methyl-2-phenyl, 2-phenyl-5-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylicacid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazol-4-ylcarboxylic acid
5-methyl-2-phenyl-1,3-oxazole-4-carboxylic acid
SDS Documents

Documents

Promotions

Promotions