missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 18735-74-5 |
---|---|
Molecular Formula | C11H8NO3 |
Molecular Weight (g/mol) | 202.19 |
MDL Number | MFCD00275475 |
InChI Key | YABCPNYCFFUVNM-UHFFFAOYSA-M |
Synonym | 5-methyl-2-phenyloxazole-4-carboxylic acid, 2-phenyl-5-methyloxazole-4-carboxylic acid, 5-methyl-2-phenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazole-4-carboxylicacid, 4-carboxy-5-methyl-2-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid,5-methyl-2-phenyl, 2-phenyl-5-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylicacid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazol-4-ylcarboxylic acid |
PubChem CID | 318533 |
IUPAC Name | 5-methyl-2-phenyl-1,3-oxazole-4-carboxylic acid |
SMILES | CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC26701CB
|
Thermo Scientific Chemicals
CC26701CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC26701DA
|
Thermo Scientific Chemicals
CC26701DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC26701DE
|
Thermo Scientific Chemicals
CC26701DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
18735-74-5 | |
202.19 | |
YABCPNYCFFUVNM-UHFFFAOYSA-M | |
318533 | |
CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O |
C11H8NO3 | |
MFCD00275475 | |
5-methyl-2-phenyloxazole-4-carboxylic acid, 2-phenyl-5-methyloxazole-4-carboxylic acid, 5-methyl-2-phenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazole-4-carboxylicacid, 4-carboxy-5-methyl-2-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid,5-methyl-2-phenyl, 2-phenyl-5-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylicacid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazol-4-ylcarboxylic acid | |
5-methyl-2-phenyl-1,3-oxazole-4-carboxylic acid |
Specifications
18735-74-5 | |
Amber Glass Bottle | |
MFCD00275475 | |
5-methyl-2-phenyloxazole-4-carboxylic acid, 2-phenyl-5-methyloxazole-4-carboxylic acid, 5-methyl-2-phenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazole-4-carboxylicacid, 4-carboxy-5-methyl-2-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid,5-methyl-2-phenyl, 2-phenyl-5-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylicacid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazol-4-ylcarboxylic acid | |
CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O | |
202.19 | |
203.2 | |
5-Methyl-2-phenyl-1, 3-oxazole-4-carboxylic acid |
97% | |
C11H8NO3 | |
250mg | |
YABCPNYCFFUVNM-UHFFFAOYSA-M | |
5-methyl-2-phenyl-1,3-oxazole-4-carboxylic acid | |
318533 | |
97% |