missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC16201DA

 View more versions of this product

Catalog No. CC16201DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

5-Methyl-2-phenyl-2H-1, 2, 3-triazole-4-carboxylic acid
96.5
97%
C10H9N3O2
1g
GQYQHAGPALBYDO-UHFFFAOYSA-N
5-methyl-2-phenyltriazole-4-carboxylic acid
519936
97%
22300-56-7
100.0
Amber Glass Bottle
MFCD00051653
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-2h-1,2,3-triazole-5-carboxylic acid, 2h-1,2,3-triazole-4-carboxylic acid, 5-methyl-2-phenyl, acmc-20ame3, maybridge1_002457, 4-carboxy-5-methyl-2-phenyl-1,2,3-triazole, 2-phenyl-5-methyl-4-carboxy-2h-1,2,3-triazole, 2h-1,2,3-triazole-4-carboxylicacid,5-methyl-2-phenyl
CC1=NN(N=C1C(=O)O)C2=CC=CC=C2
203.201
203.2

Chemical Identifiers

22300-56-7
203.201
GQYQHAGPALBYDO-UHFFFAOYSA-N
519936
CC1=NN(N=C1C(=O)O)C2=CC=CC=C2
C10H9N3O2
MFCD00051653
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-2h-1,2,3-triazole-5-carboxylic acid, 2h-1,2,3-triazole-4-carboxylic acid, 5-methyl-2-phenyl, acmc-20ame3, maybridge1_002457, 4-carboxy-5-methyl-2-phenyl-1,2,3-triazole, 2-phenyl-5-methyl-4-carboxy-2h-1,2,3-triazole, 2h-1,2,3-triazole-4-carboxylicacid,5-methyl-2-phenyl
5-methyl-2-phenyltriazole-4-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions