missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS
22300-56-7
Molecular Formula
C10H9N3O2
Molecular Weight (g/mol)
203.201
MDL Number
MFCD00051653
InChI Key
GQYQHAGPALBYDO-UHFFFAOYSA-N
Synonym
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-2h-1,2,3-triazole-5-carboxylic acid, 2h-1,2,3-triazole-4-carboxylic acid, 5-methyl-2-phenyl, acmc-20ame3, maybridge1_002457, 4-carboxy-5-methyl-2-phenyl-1,2,3-triazole, 2-phenyl-5-methyl-4-carboxy-2h-1,2,3-triazole, 2h-1,2,3-triazole-4-carboxylicacid,5-methyl-2-phenyl
PubChem CID
519936
IUPAC Name
5-methyl-2-phenyltriazole-4-carboxylic acid
SMILES
CC1=NN(N=C1C(=O)O)C2=CC=CC=C2
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
CAS
22300-56-7
Molecular Weight (g/mol)
203.201
InChI Key
GQYQHAGPALBYDO-UHFFFAOYSA-N
PubChem CID
519936
SMILES
CC1=NN(N=C1C(=O)O)C2=CC=CC=C2
Molecular Formula
C10H9N3O2
MDL Number
MFCD00051653
Synonym
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-2h-1,2,3-triazole-5-carboxylic acid, 2h-1,2,3-triazole-4-carboxylic acid, 5-methyl-2-phenyl, acmc-20ame3, maybridge1_002457, 4-carboxy-5-methyl-2-phenyl-1,2,3-triazole, 2-phenyl-5-methyl-4-carboxy-2h-1,2,3-triazole, 2h-1,2,3-triazole-4-carboxylicacid,5-methyl-2-phenyl
IUPAC Name
5-methyl-2-phenyltriazole-4-carboxylic acid
Specifications
CAS
22300-56-7
CAS Max %
100.0
Packaging
Amber Glass Bottle
MDL Number
MFCD00051653
Synonym
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-2h-1,2,3-triazole-5-carboxylic acid, 2h-1,2,3-triazole-4-carboxylic acid, 5-methyl-2-phenyl, acmc-20ame3, maybridge1_002457, 4-carboxy-5-methyl-2-phenyl-1,2,3-triazole, 2-phenyl-5-methyl-4-carboxy-2h-1,2,3-triazole, 2h-1,2,3-triazole-4-carboxylicacid,5-methyl-2-phenyl
SMILES
CC1=NN(N=C1C(=O)O)C2=CC=CC=C2
Molecular Weight (g/mol)
203.201
Formula Weight
203.2
Chemical Name or Material
5-Methyl-2-phenyl-2H-1, 2, 3-triazole-4-carboxylic acid
CAS Min %
96.5
Assay Percent Range
97%
Molecular Formula
C10H9N3O2
Quantity
1g
InChI Key
GQYQHAGPALBYDO-UHFFFAOYSA-N
IUPAC Name
5-methyl-2-phenyltriazole-4-carboxylic acid
PubChem CID
519936
Percent Purity
97%
Videos
Safety and Handling
Safety and Handling
GHS H Statement: Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'