missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(5-Methylisoxazol-3-yl)methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 35166-33-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00085129
InChI Key MDYHWQQHEWDJKR-UHFFFAOYSA-N
Synonym 5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
PubChem CID 2736567
IUPAC Name (5-methyl-1,2-oxazol-3-yl)methanol
SMILES CC1=CC(CO)=NO1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC10409DA
SDS View Documents
Thermo Scientific Chemicals
CC10409DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC10409DE
SDS View Documents
Thermo Scientific Chemicals
CC10409DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC10409EA
SDS View Documents
Thermo Scientific Chemicals
CC10409EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

35166-33-7
113.12
MDYHWQQHEWDJKR-UHFFFAOYSA-N
2736567
CC1=CC(CO)=NO1
C5H7NO2
MFCD00085129
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
(5-methyl-1,2-oxazol-3-yl)methanol

Specifications

35166-33-7
100.0
Amber glass bottle
MFCD00085129
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
CC1=CC(CO)=NO1
113.12
113.12
(5-Methylisoxazol-3-yl)methanol
96.5
97%
C5H7NO2
1g
MDYHWQQHEWDJKR-UHFFFAOYSA-N
(5-methyl-1,2-oxazol-3-yl)methanol
2736567
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'