missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nitropyridine-2-carboxylic acid, ≥97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6449906

 View more versions of this product

Catalog No. AAH6449906


Explore available promotions

Description

Description

It plays an important role in synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role in synthetic intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

5-Nitropyridine-2-carboxylic acid
208°C to 212°C
MFCD04114182
139209
Soluble in dimethyl sulfoxide.
OC(=O)C1=NC=C(C=C1)[N+]([O-])=O
168.11
168.11
Solid
30651-24-2
C6H4N2O4
5 g
5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid
QKYRCTVBMNXTBT-UHFFFAOYSA-N
5-nitropyridine-2-carboxylic acid
520488
≥97%

Chemical Identifiers

30651-24-2
168.11
QKYRCTVBMNXTBT-UHFFFAOYSA-N
520488
OC(=O)C1=NC=C(C=C1)[N+]([O-])=O
C6H4N2O4
MFCD04114182
5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid
5-nitropyridine-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only