Learn More
5-Nitropyridine-2-carboxylic acid, ≥97%, Thermo Scientific Chemicals
$66.29 - $208.18
Chemical Identifiers
CAS | 30651-24-2 |
---|---|
Molecular Formula | C6H4N2O4 |
Molecular Weight (g/mol) | 168.11 |
MDL Number | MFCD04114182 |
InChI Key | QKYRCTVBMNXTBT-UHFFFAOYSA-N |
Synonym | 5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid |
PubChem CID | 520488 |
IUPAC Name | 5-nitropyridine-2-carboxylic acid |
SMILES | OC(=O)C1=NC=C(C=C1)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6449903
|
Thermo Scientific Chemicals
H6449903 |
1 g |
Each for $66.29
|
|
|||||
AAH6449906
|
Thermo Scientific Chemicals
H6449906 |
5 g |
Each for $208.18
|
|
|||||
Description
It plays an important role in synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt plays an important role in synthetic intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
30651-24-2 | |
168.11 | |
QKYRCTVBMNXTBT-UHFFFAOYSA-N | |
520488 | |
OC(=O)C1=NC=C(C=C1)[N+]([O-])=O |
C6H4N2O4 | |
MFCD04114182 | |
5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid | |
5-nitropyridine-2-carboxylic acid |
Specifications
30651-24-2 | |
C6H4N2O4 | |
1 g | |
5-nitropicolinic acid, 5-nitropyridine-2-carboxylic acid, 5-nitro-2-pyridinecarboxylic acid, 5-nitro-2-picolinic acid, 2-pyridinecarboxylic acid, 5-nitro, picolinic acid, 5-nitro, 3-nitro-6-pyridine carboxylic acid, 2-carboxy-5-nitropyridine, 5-nitropyridine-2-carboxylicacid | |
QKYRCTVBMNXTBT-UHFFFAOYSA-N | |
5-nitropyridine-2-carboxylic acid | |
520488 | |
≥97% | |
5-Nitropyridine-2-carboxylic acid |
208°C to 212°C | |
MFCD04114182 | |
139209 | |
Soluble in dimethyl sulfoxide. | |
OC(=O)C1=NC=C(C=C1)[N+]([O-])=O | |
168.11 | |
168.11 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only