missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol 98.0+%, TCI America™
Supplier: TCI America B28665G
Specifications
| 5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol | |
| White-Yellow | |
| 5 g | |
| TUGCKICZDBMMPM-UHFFFAOYSA-N | |
| [3-[tert-butyl(diphenyl)silyl]oxy-5-(hydroxymethyl)phenyl]methanol | |
| 44629826 | |
| ≥98.0% (HPLC) |
| 482627-84-9 | |
| C24H28O3Si | |
| [3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane | |
| CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC3=CC(=CC(=C3)CO)CO | |
| 392.57 | |
| 392.57 | |
| Crystalline Powder |
Chemical Identifiers
| 482627-84-9 | |
| 392.57 | |
| [3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane | |
| [3-[tert-butyl(diphenyl)silyl]oxy-5-(hydroxymethyl)phenyl]methanol |
| C24H28O3Si | |
| TUGCKICZDBMMPM-UHFFFAOYSA-N | |
| 44629826 | |
| CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC3=CC(=CC(=C3)CO)CO |
Safety and Handling
TSCA : No