missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol 98.0+%, TCI America™
$683.35 - $3217.27
Chemical Identifiers
CAS | 482627-84-9 |
---|---|
Molecular Formula | C24H28O3Si |
Molecular Weight (g/mol) | 392.57 |
InChI Key | TUGCKICZDBMMPM-UHFFFAOYSA-N |
Synonym | [3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane |
PubChem CID | 44629826 |
IUPAC Name | [3-[tert-butyl(diphenyl)silyl]oxy-5-(hydroxymethyl)phenyl]methanol |
SMILES | CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC3=CC(=CC(=C3)CO)CO |
Chemical Identifiers
482627-84-9 | |
392.57 | |
[3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane | |
[3-[tert-butyl(diphenyl)silyl]oxy-5-(hydroxymethyl)phenyl]methanol |
C24H28O3Si | |
TUGCKICZDBMMPM-UHFFFAOYSA-N | |
44629826 | |
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC3=CC(=CC(=C3)CO)CO |
Specifications
482627-84-9 | |
C24H28O3Si | |
[3,5-Bis(hydroxymethyl)phenoxy]-tert-butyldiphenylsilane | |
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC3=CC(=CC(=C3)CO)CO | |
392.57 | |
392.57 | |
Crystalline Powder |
White-Yellow | |
1 g | |
TUGCKICZDBMMPM-UHFFFAOYSA-N | |
[3-[tert-butyl(diphenyl)silyl]oxy-5-(hydroxymethyl)phenyl]methanol | |
44629826 | |
≥98.0% (HPLC) | |
5-(tert-Butyldiphenylsilyloxy)-1,3-benzenedimethanol |
Safety and Handling
TSCA : No