3M™ 5000 Series Disposable Half-Facepiece Respirators
Preassembled respirators are maintenance-free and require no spare parts
Supplier:  3M™ 7000051848
 View more versions of this product
Catalog No. 17-676C
Explore available promotions
Includes: Organic vapor cartridge and reusable storage bag.
Description
Description
  • Ideally suited for intermittent use, plant shutdowns, and applications where respirators wear out or become difficult to clean in short periods of time
  • Color coded for easy size identification
  • Can be used with NIOSH-approved 3M filters N95, P95, and P100 for oil and non-oil particles
  • For convenience and simplicity, some respirator assemblies are packaged with 3M Filters P95 or P100 along with 3M Filter Retainer 501 or Filter Adapter 502
  • Some styles feature acid gas and/or organic vapor protection
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
NIOSH 42 CFR Part 84
Thermoplastic Elastomer
Large
Assembly, cleaning, grinding, machine operation, painting, welding
Organic vapor cartridge, reusable storage bag
4-point
0.22 kg
Yellow
Half-Mask Respirator
Thermoplastic Elastomer
3M filters N95, P95, and P100 for oil and non-oil particles
5303
0.5 lb.