missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ 5000 Series Disposable Half-Facepiece Respirators

Preassembled respirators are maintenance-free and require no spare parts

$47.92 - $499.41

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) 3M filters N95, P95, and P100 for oil and non-oil particles
View More Specs

Products 7
Catalog Number Mfr. No. Color Size For Use With (Application) Includes Price Quantity  
Catalog Number Mfr. No. Color Size For Use With (Application) Includes Price Quantity  
17-674B
View Documents
3M™
7000126026
Black Medium Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
Case of 12 Each for $421.44
 
17-674C
View Documents
3M™
7000126027
Black Large Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
Case of 12 Each for $470.95
 
17-676A
View Documents
3M™
7000126025
Yellow Small Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
Case of 12 Each for $446.91
 
17-676B
View Documents
3M™
7000051847
Yellow Medium Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
Case of 12 Each for $499.41
 
17-676C
View Documents
3M™
7000051848
Yellow Large Assembly, cleaning, grinding, machine operation, painting, welding Organic vapor cartridge, reusable storage bag
Case of 12 Each for $446.91
 
19-003453A
View Documents
3M™
7000052079
Black Small Spray painting, pesticides, agriculture, construction and transportation Organic vapor cartridge, reusable storage bag
Each for $55.34
Case of 12 Each for $205.92
 
19-003453B
View Documents
3M™
7000052080
Black Medium Spray painting, pesticides, agriculture, construction and transportation Organic vapor cartridge, reusable storage bag
Each for $47.92
Case of 12 Each for $532.20
 
Description

Description

  • Ideally suited for intermittent use, plant shutdowns, and applications where respirators wear out or become difficult to clean in short periods of time
  • Color coded for easy size identification
  • Can be used with NIOSH-approved 3M filters N95, P95, and P100 for oil and non-oil particles
  • For convenience and simplicity, some respirator assemblies are packaged with 3M Filters P95 or P100 along with 3M Filter Retainer 501 or Filter Adapter 502
  • Some styles feature acid gas and/or organic vapor protection

Certifications

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Compliance

NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.

Specifications

Specifications

NIOSH 42 CFR Part 84
Half-Mask Respirator
3M filters N95, P95, and P100 for oil and non-oil particles
4-point
Thermoplastic Elastomer
Thermoplastic Elastomer
Organic vapor cartridge, reusable storage bag
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'