National Safety Apparel™ 55 Cal ArcGuard™ TECGEN FR Arc Flash Kit with Gloves (10)
Kit includes: 55 cal TECGEN CC short coat and bib overall, Arc flash hood with PureView faceshield and hard hat, safety glasses, hood bag and gear bag
Supplier: National Safety Apparel™ KIT4SC55MD10
Description
- Arc Rating: 55 cal/cm²
- CAT 4 Fabric/Material: Double-layer TECGEN CC™
- 30% lighter than standard 40 cal kits
- FR hook and loop front closure
- Stand up collar for extra coverage
- Rib knit cuffs for comfortable, secure fit
- Garments embroidered with arc rating
- Quick disconnect and adjustable shoulder straps on bib overalls
- Expandable leg opening to fit over boots
- Hook and loop at side of bib for easy access
- Made in the USA
Specifications
55 cal/cm² | |
CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269 | |
Double-Layer TECGEN CC™ | |
FR Hook and Loop | |
Adjustable |
Navy | |
Open | |
Medium | |
Stitched |