National Safety Apparel™ 55 Cal ArcGuard™ TECGEN FR Arc Flash Kit with Gloves (10)
Kit includes: 55 cal TECGEN CC short coat and bib overall, Arc flash hood with PureView faceshield and hard hat, safety glasses, hood bag and gear bag
Supplier:  National Safety Apparel™ KIT4SC554X10
 View more versions of this product
Catalog No. 170011793
Explore available promotions
Description
Description
  • Arc Rating: 55 cal/cm²
  • CAT 4 Fabric/Material: Double-layer TECGEN CC™
  • 30% lighter than standard 40 cal kits
  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Hook and loop at side of bib for easy access
  • Made in the USA
Specifications
Specifications
55 cal/cm²
CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Double-Layer TECGEN CC™
FR Hook and Loop
Adjustable
Navy
Open
4X-Large
Stitched