missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6,9-Dichloro-2-methoxyacridine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0591314

 View more versions of this product

Catalog No. AAL0591314


Explore available promotions

Description

Description

6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N'-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N'-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N′-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N′-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

6,9-Dichloro-2-methoxyacridine
163°C to 165°C
MFCD00005028
226309
Insoluble in water.
COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl
278.132
278.14
86-38-4
C14H9Cl2NO
25 g
halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine
RYRNQWYNHLLOGX-UHFFFAOYSA-N
6,9-dichloro-2-methoxyacridine
66577
98%

Chemical Identifiers

86-38-4
278.132
RYRNQWYNHLLOGX-UHFFFAOYSA-N
66577
COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl
C14H9Cl2NO
MFCD00005028
halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine
6,9-dichloro-2-methoxyacridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

EINECSNumber : 201-666-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only