Learn More
6,9-Dichloro-2-methoxyacridine, 98%, Thermo Scientific Chemicals
$71.49 - $210.96
Chemical Identifiers
CAS | 86-38-4 |
---|---|
Molecular Formula | C14H9Cl2NO |
Molecular Weight (g/mol) | 278.132 |
MDL Number | MFCD00005028 |
InChI Key | RYRNQWYNHLLOGX-UHFFFAOYSA-N |
Synonym | halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine |
PubChem CID | 66577 |
IUPAC Name | 6,9-dichloro-2-methoxyacridine |
SMILES | COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0591306
|
Thermo Scientific Chemicals
L0591306 |
5 g |
Each for $71.49
|
|
|||||
AAL0591314
|
Thermo Scientific Chemicals
L0591314 |
25 g |
Each for $210.96
|
|
|||||
Description
6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N'-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N'-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N′-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N′-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
86-38-4 | |
278.132 | |
RYRNQWYNHLLOGX-UHFFFAOYSA-N | |
66577 | |
COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl |
C14H9Cl2NO | |
MFCD00005028 | |
halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine | |
6,9-dichloro-2-methoxyacridine |
Specifications
86-38-4 | |
C14H9Cl2NO | |
5 g | |
halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine | |
RYRNQWYNHLLOGX-UHFFFAOYSA-N | |
6,9-dichloro-2-methoxyacridine | |
66577 | |
98% |
163°C to 165°C | |
MFCD00005028 | |
226309 | |
Insoluble in water. | |
COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl | |
278.132 | |
278.14 | |
6,9-Dichloro-2-methoxyacridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
EINECSNumber : 201-666-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only