missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1466614

 View more versions of this product

Catalog No. AAA1466614


Description

Description

6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Product is sensitive to light and moisture. Handle and store under inert gas. Keep away from strong oxidizing agents.
Specifications

Specifications

6-Acetyl-1,2,3,4-tetrahydronaphthalene
-21°C
119°C to 121°C (2 mmHg)
C12H14O
MFCD00019710
1241458
Slightly soluble in water.
CC(=O)C1=CC=C2CCCCC2=C1
174.24
174.24
774-55-0
1.058
>110°C (230°F)
1.561
25 g
1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one
VEPUKHYQNXSSKV-UHFFFAOYSA-N
1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethanone
69885
97%

Chemical Identifiers

774-55-0
174.24
VEPUKHYQNXSSKV-UHFFFAOYSA-N
69885
C12H14O
MFCD00019710
1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one
CC(=O)C1=CC=C2CCCCC2=C1
Safety and Handling

Safety and Handling

EINECSNumber : 212-266-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only