Learn More
6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals
$85.04 - $748.52
Chemical Identifiers
CAS | 774-55-0 |
---|---|
Molecular Formula | C12H14O |
Molecular Weight (g/mol) | 174.24 |
MDL Number | MFCD00019710 |
InChI Key | VEPUKHYQNXSSKV-UHFFFAOYSA-N |
Synonym | 1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one |
PubChem CID | 69885 |
SMILES | CC(=O)C1=CC=C2CCCCC2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1466606
|
Thermo Scientific Chemicals
A1466606 |
5 g |
Each for $85.04
|
|
|||||
AAA1466614
|
Thermo Scientific Chemicals
A1466614 |
25 g |
Each for $255.81
|
|
|||||
AAA1466622
|
Thermo Scientific Chemicals
A1466622 |
100 g |
Each for $748.52
|
|
|||||
Description
6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Product is sensitive to light and moisture. Handle and store under inert gas. Keep away from strong oxidizing agents.
Chemical Identifiers
774-55-0 | |
174.24 | |
VEPUKHYQNXSSKV-UHFFFAOYSA-N | |
69885 |
C12H14O | |
MFCD00019710 | |
1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one | |
CC(=O)C1=CC=C2CCCCC2=C1 |
Specifications
774-55-0 | |
1.058 | |
>110°C (230°F) | |
1.561 | |
5 g | |
1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one | |
VEPUKHYQNXSSKV-UHFFFAOYSA-N | |
1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethanone | |
69885 | |
97% |
-21°C | |
119°C to 121°C (2 mmHg) | |
C12H14O | |
MFCD00019710 | |
1241458 | |
Slightly soluble in water. | |
CC(=O)C1=CC=C2CCCCC2=C1 | |
174.24 | |
174.24 | |
6-Acetyl-1,2,3,4-tetrahydronaphthalene |
Safety and Handling
EINECSNumber : 212-266-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only