Learn More
6-Acetyl-1,2,3,4-tetrahydronaphthalene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1466622
Description
6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Acetyl-1,2,3,4-tetrahydronaphthalene, is used asa novel methyl anthranilate yet extremely stable and resistant to discoloration. It is used in liquid detergent, concentrated fabric softner, alcoholic fine fragrances, antiperspirants. It is also used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Product is sensitive to light and moisture. Handle and store under inert gas. Keep away from strong oxidizing agents.
Specifications
6-Acetyl-1,2,3,4-tetrahydronaphthalene | |
-21°C | |
119°C to 121°C (2 mmHg) | |
C12H14O | |
MFCD00019710 | |
1241458 | |
Slightly soluble in water. | |
CC(=O)C1=CC=C2CCCCC2=C1 | |
174.24 | |
174.24 |
774-55-0 | |
1.058 | |
>110°C (230°F) | |
1.561 | |
100 g | |
1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one | |
VEPUKHYQNXSSKV-UHFFFAOYSA-N | |
1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one | |
69885 | |
97% |
Chemical Identifiers
774-55-0 | |
174.24 | |
VEPUKHYQNXSSKV-UHFFFAOYSA-N | |
69885 |
C12H14O | |
MFCD00019710 | |
1-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 6-acetyltetralin, 6-acetyl tetralin, ethanone, 1-5,6,7,8-tetrahydro-2-naphthalenyl, unii-6l3dbr6de6, 6-acetyl-1,2,3,4-tetrahydronaphthalene, 5,6,7,8-tetrahydro-2-acetonaphthone, 6l3dbr6de6, 6-acetyltetrahydronaphthalene, 1-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one | |
CC(=O)C1=CC=C2CCCCC2=C1 |
Safety and Handling
EINECSNumber : 212-266-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only