missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Aminocaproic acid, 99+%, Thermo Scientific Chemicals

$161.13 - $161.13

Chemical Identifiers

CAS 60-32-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.17
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
PubChem CID 564
ChEBI CHEBI:16586
IUPAC Name 6-aminohexanoic acid
SMILES C(CCC(=O)O)CCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC103301000
SDS View Documents
Thermo Scientific Chemicals
103301000
100 g Plastic bottle
Each for $161.13
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60-32-2
131.17
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
CHEBI:16586
C(CCC(=O)O)CCN
C6H13NO2
SLXKOJJOQWFEFD-UHFFFAOYSA-N
564
6-aminohexanoic acid

Specifications

60-32-2
White
99+%
C6H13NO2
100 g
15,427
(13g/100ml H2O) Clear colorless solution
C(CCC(=O)O)CCN
131.17
CHEBI:16586
99+%
6-Aminocaproic acid
204.0°C to 209.0°C
Authentic
Plastic bottle
H2N(CH2)5CO2H
04,434
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
SLXKOJJOQWFEFD-UHFFFAOYSA-N
6-aminohexanoic acid
564
131.17
Crystalline Powder
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 200-469-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only