missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-(Boc-amino)hexanoic acid, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6135014

 View more versions of this product

Catalog No. AAH6135014

Explore available promotions

Description

Description

6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.

Solubility
Soluble in chloroform and ethyl acetate. Slightly soluble in water.

Notes
Store at -20°C. Keep container tightly closed. With good ventilation. Protect form heat and store away from oxidizing agent.
Specifications

Specifications

6-(Boc-amino)hexanoic acid
35°C to 40°C
C11H21NO4
MFCD00037798
2049561
Soluble in chloroform and ethyl acetate. Slightly soluble in water.
CC(C)(C)OC(=O)NCCCCCC(=O)O
231.292
231.29
Powder
6404-29-1
White
(CH3)3COCONH(CH2)5CO2H
25 g
boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid
RUFDYIJGNPVTAY-UHFFFAOYSA-N
6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
637602
95%

Chemical Identifiers

6404-29-1
231.292
RUFDYIJGNPVTAY-UHFFFAOYSA-N
637602
CC(C)(C)OC(=O)NCCCCCC(=O)O
C11H21NO4
MFCD00037798
boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid
6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only