Learn More
6-(Boc-amino)hexanoic acid, 95%, Thermo Scientific Chemicals
$142.99 - $562.47
Chemical Identifiers
CAS | 6404-29-1 |
---|---|
Molecular Formula | C11H21NO4 |
Molecular Weight (g/mol) | 231.292 |
MDL Number | MFCD00037798 |
InChI Key | RUFDYIJGNPVTAY-UHFFFAOYSA-N |
Synonym | boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid |
PubChem CID | 637602 |
IUPAC Name | 6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
SMILES | CC(C)(C)OC(=O)NCCCCCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6135006
|
Thermo Scientific Chemicals
H6135006 |
5 g |
N/A
|
|
||||||
AAH6135014
|
Thermo Scientific Chemicals
H6135014 |
25 g |
N/A
|
|
||||||
Description
6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-(Boc-amino)hexanoic acid is used in the preparation of esters of 6-aminohexanoic acid as antibacterial agents. EACA is reported to inhibit chymotrypsin, Factor VIIa, lysine carboxy peptidase, plasmin, and plasminogen activator.
Solubility
Soluble in chloroform and ethyl acetate. Slightly soluble in water.
Notes
Store at -20°C. Keep container tightly closed. With good ventilation. Protect form heat and store away from oxidizing agent.
Chemical Identifiers
6404-29-1 | |
231.292 | |
RUFDYIJGNPVTAY-UHFFFAOYSA-N | |
637602 | |
CC(C)(C)OC(=O)NCCCCCC(=O)O |
C11H21NO4 | |
MFCD00037798 | |
boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid | |
6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Specifications
6404-29-1 | |
White | |
(CH3)3COCONH(CH2)5CO2H | |
5 g | |
boc-epsilon-acp-oh, boc-6-aminocaproic acid, 6-tert-butoxycarbonyl amino hexanoic acid, boc-6-ahx-oh, 6-boc-amino hexanoic acid, boc-, a-acp-oh, hexanoic acid, 6-1,1-dimethylethoxy carbonyl amino, n-boc-6-aminohexanoic acid, 6-boc-amino caproic acid | |
RUFDYIJGNPVTAY-UHFFFAOYSA-N | |
6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
637602 | |
95% | |
6-(Boc-amino)hexanoic acid |
35°C to 40°C | |
C11H21NO4 | |
MFCD00037798 | |
2049561 | |
Soluble in chloroform and ethyl acetate. Slightly soluble in water. | |
CC(C)(C)OC(=O)NCCCCCC(=O)O | |
231.292 | |
231.29 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only