missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-2,3-difluorophenylacetonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H3329603

 View more versions of this product

Catalog No. AAH3329603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

6-Chloro-2,3-difluorophenylacetonitrile
C8H4ClF2N
1g
6-chloro-2,3-difluorophenylacetonitrile, 2-6-chloro-2,3-difluorophenyl acetonitrile
C1=CC(=C(C(=C1F)F)CC#N)Cl
187.574
187.57
1373920-84-3
MFCD22201041
UN3439
DZNGMYTZQJRVAF-UHFFFAOYSA-N
2-(6-chloro-2,3-difluorophenyl)acetonitrile
86277643
97%

Chemical Identifiers

1373920-84-3
187.574
DZNGMYTZQJRVAF-UHFFFAOYSA-N
86277643
C1=CC(=C(C(=C1F)F)CC#N)Cl
C8H4ClF2N
MFCD22201041
6-chloro-2,3-difluorophenylacetonitrile, 2-6-chloro-2,3-difluorophenyl acetonitrile
2-(6-chloro-2,3-difluorophenyl)acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions