missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Chloro-2,3-difluorophenylacetonitrile, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1373920-84-3
Molecular Formula C8H4ClF2N
Molecular Weight (g/mol) 187.574
MDL Number MFCD22201041
InChI Key DZNGMYTZQJRVAF-UHFFFAOYSA-N
Synonym 6-chloro-2,3-difluorophenylacetonitrile, 2-6-chloro-2,3-difluorophenyl acetonitrile
PubChem CID 86277643
IUPAC Name 2-(6-chloro-2,3-difluorophenyl)acetonitrile
SMILES C1=CC(=C(C(=C1F)F)CC#N)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3329603
View Documents
Thermo Scientific Chemicals
H3329603
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1373920-84-3
187.574
DZNGMYTZQJRVAF-UHFFFAOYSA-N
86277643
C1=CC(=C(C(=C1F)F)CC#N)Cl
C8H4ClF2N
MFCD22201041
6-chloro-2,3-difluorophenylacetonitrile, 2-6-chloro-2,3-difluorophenyl acetonitrile
2-(6-chloro-2,3-difluorophenyl)acetonitrile

Specifications

1373920-84-3
MFCD22201041
UN3439
DZNGMYTZQJRVAF-UHFFFAOYSA-N
2-(6-chloro-2,3-difluorophenyl)acetonitrile
86277643
97%
C8H4ClF2N
1g
6-chloro-2,3-difluorophenylacetonitrile, 2-6-chloro-2,3-difluorophenyl acetonitrile
C1=CC(=C(C(=C1F)F)CC#N)Cl
187.574
187.57
6-Chloro-2,3-difluorophenylacetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'