missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hepten-1-ol 96.0+%, TCI America™

Supplier:  TCI America H094925G

 View more versions of this product

Catalog No. H094925G


Explore available promotions

Description

Description

Specifications

Specifications

6-Hepten-1-ol
Colorless
C7H14O
25 g
UFULDTPDHIRNGS-UHFFFAOYSA-N
hept-6-en-1-ol
543123
≥96.0% (GC)
4117-10-6
113°C
MFCD00229167
1987
C=CCCCCCO
114.19
114.19
Liquid

Chemical Identifiers

4117-10-6
114.19
UFULDTPDHIRNGS-UHFFFAOYSA-N
hept-6-en-1-ol
C7H14O
MFCD00229167
543123
C=CCCCCCO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

TSCA : No

SDS
Documents

Documents

Promotions

Promotions