missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

6-Hepten-1-ol 96.0+%, TCI America™

$817.89 - $3271.56

Chemical Identifiers

CAS 4117-10-6
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00229167
InChI Key UFULDTPDHIRNGS-UHFFFAOYSA-N
PubChem CID 543123
IUPAC Name hept-6-en-1-ol
SMILES C=CCCCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09495G
View Documents Promotion Details
TCI America
H09495G
5 g
Each for $817.89
 
H094925G
View Documents Promotion Details
TCI America
H094925G
25 g
Each for $3,271.56
 
Description

Description

Specifications

Chemical Identifiers

4117-10-6
114.19
UFULDTPDHIRNGS-UHFFFAOYSA-N
hept-6-en-1-ol
C7H14O
MFCD00229167
543123
C=CCCCCCO

Specifications

4117-10-6
113°C
MFCD00229167
1987
C=CCCCCCO
114.19
114.19
Liquid
Colorless
C7H14O
5 g
UFULDTPDHIRNGS-UHFFFAOYSA-N
hept-6-en-1-ol
543123
≥96.0% (GC)
6-Hepten-1-ol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'