missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Mercapto-1-hexanol 98.0+%, TCI America™
$159.27 - $630.41
Chemical Identifiers
CAS | 1633-78-9 |
---|---|
Molecular Formula | C6H14OS |
Molecular Weight (g/mol) | 134.237 |
MDL Number | MFCD00068552 |
InChI Key | UGZAJZLUKVKCBM-UHFFFAOYSA-N |
PubChem CID | 560126 |
IUPAC Name | 6-sulfanylhexan-1-ol |
SMILES | C(CCCS)CCO |
Chemical Identifiers
1633-78-9 | |
134.237 | |
UGZAJZLUKVKCBM-UHFFFAOYSA-N | |
6-sulfanylhexan-1-ol |
C6H14OS | |
MFCD00068552 | |
560126 | |
C(CCCS)CCO |
Specifications
1633-78-9 | |
105°C | |
MFCD00068552 | |
UGZAJZLUKVKCBM-UHFFFAOYSA-N | |
6-sulfanylhexan-1-ol | |
560126 | |
≥98.0% (GC) | |
6-Mercapto-1-hexanol |
Colorless | |
C6H14OS | |
5 g | |
C(CCCS)CCO | |
134.237 | |
134.24 | |
Liquid |
Safety and Handling
TSCA : No