Learn More
6-Methylindole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 133880010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
6-Methylindole | |
98.5 | |
Yellow to Brown | |
112°C (5.0 mmHg) | |
Authentic | |
Glass bottle | |
1.6060 to 1.608 | |
1 g | |
1.05 | |
ONYNOPPOVKYGRS-UHFFFAOYSA-N | |
tert-butyl N-phenylcarbamate | |
137930 | |
99% |
3420-02-8 | |
100.0 | |
1.0500g/mL | |
>112°C | |
98.5% min. (GC) | |
C9H9N | |
MFCD00005682 | |
20, IV, 3214 | |
n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester | |
CC1=CC=C2C=CNC2=C1 | |
131.18 | |
131.18 | |
Liquid After Melting |
Chemical Identifiers
3420-02-8 | |
131.18 | |
ONYNOPPOVKYGRS-UHFFFAOYSA-N | |
137930 |
C9H9N | |
MFCD00005682 | |
n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester | |
CC1=CC=C2C=CNC2=C1 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Recommended Storage : This product darkens on exposure to light
RUO – Research Use Only