missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Trifluoromethyl-4-pyrimidinol 98.0+%, TCI America™
$60.57 - $217.06
Chemical Identifiers
CAS | 1546-78-7 |
---|---|
Molecular Formula | C5H3F3N2O |
Molecular Weight (g/mol) | 164.087 |
MDL Number | MFCD00192530 |
InChI Key | LVOYSBZJJWPUBD-UHFFFAOYSA-N |
Synonym | 4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl |
PubChem CID | 1268071 |
IUPAC Name | 6-(trifluoromethyl)-1H-pyrimidin-4-one |
SMILES | C1=C(NC=NC1=O)C(F)(F)F |
Chemical Identifiers
1546-78-7 | |
164.087 | |
LVOYSBZJJWPUBD-UHFFFAOYSA-N | |
1268071 | |
C1=C(NC=NC1=O)C(F)(F)F |
C5H3F3N2O | |
MFCD00192530 | |
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl | |
6-(trifluoromethyl)-1H-pyrimidin-4-one |
Specifications
1546-78-7 | |
White-Yellow | |
MFCD00192530 | |
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl | |
C1=C(NC=NC1=O)C(F)(F)F | |
164.087 | |
164.09 | |
Crystalline Powder |
170°C | |
C5H3F3N2O | |
5 g | |
LVOYSBZJJWPUBD-UHFFFAOYSA-N | |
6-(trifluoromethyl)-1H-pyrimidin-4-one | |
1268071 | |
≥98.0% (GC,T) | |
6-Trifluoromethyl-4-pyrimidinol |
Safety and Handling
TSCA : No