missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
6-Trifluoromethyl-4-pyrimidinol 98.0+%, TCI America™
Supplier: TCI America T16015G
Specifications
6-Trifluoromethyl-4-pyrimidinol | |
170°C | |
C5H3F3N2O | |
5 g | |
LVOYSBZJJWPUBD-UHFFFAOYSA-N | |
6-(trifluoromethyl)-1H-pyrimidin-4-one | |
1268071 | |
≥98.0% (GC,T) |
1546-78-7 | |
White-Yellow | |
MFCD00192530 | |
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl | |
C1=C(NC=NC1=O)C(F)(F)F | |
164.087 | |
164.09 | |
Crystalline Powder |
Chemical Identifiers
1546-78-7 | |
164.087 | |
LVOYSBZJJWPUBD-UHFFFAOYSA-N | |
1268071 | |
C1=C(NC=NC1=O)C(F)(F)F |
C5H3F3N2O | |
MFCD00192530 | |
4-hydroxy-6-trifluoromethyl pyrimidine, 4-hydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidin-4-ol, 6-trifluoromethyl-4-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-4-one, 6-trifluoromethyl-pyrimidin-4-ol, 6-trifluoromethyl pyrimidin-4 3h-one, 6-trifluoromethyl-4-hydroxypyrimidine, 4 1h-pyrimidinone, 6-trifluoromethyl | |
6-(trifluoromethyl)-1H-pyrimidin-4-one |
Safety and Handling
TSCA : No