missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Bromo-1-heptene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6412714

 View more versions of this product

Catalog No. AAH6412714


Explore available promotions

Description

Description

It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

7-Bromo-1-heptene
Colorless to Yellow
27°C (1 mmHg)
C7H13Br
Br(CH2)5CH=CH2
25 g
1740861
Slightly soluble in water.
C=CCCCCCBr
177.09
177.08
Liquid
4117-09-3
1.162 g/mL
58°C (136°F)
1.466
MFCD01631137
UN1993
7-bromo-1-heptene, 1-heptene, 7-bromo, bromo-7 heptene-1, 7-bromo-heptene, 7-bromo-l-heptene, 1-bromo-6-heptene, acmc-2097en, ksc235k6r
GNYDYUQVALBGGZ-UHFFFAOYSA-N
7-bromohept-1-ene
524450
97%

Chemical Identifiers

4117-09-3
177.09
GNYDYUQVALBGGZ-UHFFFAOYSA-N
C7H13Br
MFCD01631137
7-bromohept-1-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only