Learn More
7-Bromo-1-heptene, 97%, Thermo Scientific Chemicals
$110.48 - $1577.82
Chemical Identifiers
CAS | 4117-09-3 |
---|---|
Molecular Formula | C7H13Br |
Molecular Weight (g/mol) | 177.09 |
MDL Number | MFCD01631137 |
InChI Key | GNYDYUQVALBGGZ-UHFFFAOYSA-N |
IUPAC Name | 7-bromohept-1-ene |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6412706
|
Thermo Scientific Chemicals
H6412706 |
5 g |
Each for $110.48
|
|
|||||
AAH6412714
|
Thermo Scientific Chemicals
H6412714 |
25 g |
Each for $425.49
|
|
|||||
AAH6412722
|
Thermo Scientific Chemicals
H6412722 |
100 g |
Each for $1,577.82
|
|
|||||
Description
It is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in synthetic chemistry as an alkylating agent. Also used as a reagent in the synthesis of (R)-(+)-Lasiodiplodin, a macrolide that exhibits potent antileukemic activity against human tumors.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
4117-09-3 | |
177.09 | |
GNYDYUQVALBGGZ-UHFFFAOYSA-N |
C7H13Br | |
MFCD01631137 | |
7-bromohept-1-ene |
Specifications
4117-09-3 | |
1.162 g/mL | |
58°C (136°F) | |
1.466 | |
MFCD01631137 | |
UN1993 | |
7-bromo-1-heptene, 1-heptene, 7-bromo, bromo-7 heptene-1, 7-bromo-heptene, 7-bromo-l-heptene, 1-bromo-6-heptene, acmc-2097en, ksc235k6r | |
GNYDYUQVALBGGZ-UHFFFAOYSA-N | |
7-bromohept-1-ene | |
524450 | |
97% | |
7-Bromo-1-heptene |
Colorless to Yellow | |
27°C (1 mmHg) | |
C7H13Br | |
Br(CH2)5CH=CH2 | |
5 g | |
1740861 | |
Slightly soluble in water. | |
C=CCCCCCBr | |
177.09 | |
177.08 | |
Liquid |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only