missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6103203

 View more versions of this product

Catalog No. AAH6103203

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in water (0.76 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

7-Nitro-3,4-dihydro-2H-1,4-benzoxazine
C8H8N2O3
1 g
Sparingly soluble in water (0.76g/L at 25°C).
C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]
180.163
180.16
120711-81-1
MFCD11603433
7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 2,3-dihydro-7-nitro-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine
YKCFDUNYLMTXFC-UHFFFAOYSA-N
7-nitro-3,4-dihydro-2H-1,4-benzoxazine
18416151
97%

Chemical Identifiers

120711-81-1
180.163
YKCFDUNYLMTXFC-UHFFFAOYSA-N
18416151
C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]
C8H8N2O3
MFCD11603433
7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 2,3-dihydro-7-nitro-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine
7-nitro-3,4-dihydro-2H-1,4-benzoxazine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only