missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

$367.07 - $367.07

Chemical Identifiers

CAS 120711-81-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD11603433
InChI Key YKCFDUNYLMTXFC-UHFFFAOYSA-N
Synonym 7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 2,3-dihydro-7-nitro-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine
PubChem CID 18416151
IUPAC Name 7-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAH6103203
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
H6103203
1 g
Each for $367.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Sparingly soluble in water (0.76 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

120711-81-1
180.163
YKCFDUNYLMTXFC-UHFFFAOYSA-N
18416151
C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]
C8H8N2O3
MFCD11603433
7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 2,3-dihydro-7-nitro-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine
7-nitro-3,4-dihydro-2H-1,4-benzoxazine

Specifications

120711-81-1
MFCD11603433
7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 2,3-dihydro-7-nitro-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine
YKCFDUNYLMTXFC-UHFFFAOYSA-N
7-nitro-3,4-dihydro-2H-1,4-benzoxazine
18416151
97%
C8H8N2O3
1 g
Sparingly soluble in water (0.76g/L at 25°C).
C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]
180.163
180.16
7-Nitro-3,4-dihydro-2H-1,4-benzoxazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only