missing translation for 'onlineSavingsMsg'
Learn More

8-Methylnonanoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 342340010

 View more versions of this product

Catalog No. AC342340010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

8-Methylnonanoic acid
5963-14-4
100.0
Authentic
Glass bottle
1.4330 to 1.4350
MFCD00044086
isodecanoic acid, isocapric acid, unii-lw5786qckt, 8-methyl-nonanoic acid, lw5786qckt, nonanoic acid, 8-methyl, versatic acid
OAOABCKPVCUNKO-UHFFFAOYSA-M
8-methylnonanoic acid
111470
97%
97%
96.0
118.0°C to 120.0°C (2.0 mmHg)
96% min. (GC)
C10H19O2
(CH3)2CH(CH2)6COOH
1g
Solubility in water: .. Other solubilities: soluble in thf
CC(C)CCCCCCC([O-])=O
171.26
172.27

Chemical Identifiers

5963-14-4
171.26
OAOABCKPVCUNKO-UHFFFAOYSA-M
111470
CC(C)CCCCCCC([O-])=O
C10H19O2
MFCD00044086
isodecanoic acid, isocapric acid, unii-lw5786qckt, 8-methyl-nonanoic acid, lw5786qckt, nonanoic acid, 8-methyl, versatic acid
8-methylnonanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only