missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Methylnonanoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5963-14-4
Molecular Formula C10H19O2
Molecular Weight (g/mol) 171.26
MDL Number MFCD00044086
InChI Key OAOABCKPVCUNKO-UHFFFAOYSA-M
Synonym isodecanoic acid, isocapric acid, unii-lw5786qckt, 8-methyl-nonanoic acid, lw5786qckt, nonanoic acid, 8-methyl, versatic acid
PubChem CID 111470
IUPAC Name 8-methylnonanoic acid
SMILES CC(C)CCCCCCC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC342342500
SDS View Documents
Thermo Scientific Chemicals
342342500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC342340010
SDS View Documents
Thermo Scientific Chemicals
342340010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5963-14-4
171.26
OAOABCKPVCUNKO-UHFFFAOYSA-M
111470
CC(C)CCCCCCC([O-])=O
C10H19O2
MFCD00044086
isodecanoic acid, isocapric acid, unii-lw5786qckt, 8-methyl-nonanoic acid, lw5786qckt, nonanoic acid, 8-methyl, versatic acid
8-methylnonanoic acid

Specifications

5963-14-4
100.0
Authentic
Glass bottle
1.4330 to 1.4350
MFCD00044086
isodecanoic acid, isocapric acid, unii-lw5786qckt, 8-methyl-nonanoic acid, lw5786qckt, nonanoic acid, 8-methyl, versatic acid
OAOABCKPVCUNKO-UHFFFAOYSA-M
8-methylnonanoic acid
111470
97%
96.0
118.0°C to 120.0°C (2.0 mmHg)
96% min. (GC)
C10H19O2
(CH3)2CH(CH2)6COOH
250mg
Solubility in water: .. Other solubilities: soluble in thf
CC(C)CCCCCCC([O-])=O
171.26
172.27
8-Methylnonanoic acid, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only