missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117200010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
9, 10-Diphenylanthracene | |
1499-10-1 | |
100.0 | |
Yellow to Beige | |
97.5% min. (HPLC) | |
C26H18 | |
1 g | |
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard | |
FCNCGHJSNVOIKE-UHFFFAOYSA-N | |
9,10-diphenylanthracene | |
15159 | |
330.42 | |
Fine Crystalline Powder |
99% | |
97.5 | |
248°C to 252°C | |
Authentic | |
Glass bottle | |
MFCD00001253 | |
05,747 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,chloroform,benzene,carbon,disulfide,cyclohexane,acetonitrile,petroleum,ether | |
C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 | |
330.43 | |
CHEBI:51676 | |
98% |
Chemical Identifiers
1499-10-1 | |
330.43 | |
FCNCGHJSNVOIKE-UHFFFAOYSA-N | |
15159 | |
9,10-diphenylanthracene |
C26H18 | |
MFCD00001253 | |
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard | |
CHEBI:51676 | |
C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 |
Safety and Handling
EINECSNumber : 216-105-1
TSCA : TSCA
RUO – Research Use Only