missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
9,10-Diphenylanthracene, 98%, Thermo Scientific Chemicals
$82.41 - $316.22
Identifiants chimiques
CAS | 1499-10-1 |
---|---|
Molecular Formula | C26H18 |
Molecular Weight (g/mol) | 330.43 |
MDL Number | MFCD00001253 |
InChI Key | FCNCGHJSNVOIKE-UHFFFAOYSA-N |
Synonym | anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard |
PubChem CID | 15159 |
ChEBI | CHEBI:51676 |
IUPAC Name | 9,10-diphenylanthracene |
SMILES | C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
AC117200010
|
Thermo Scientific Chemicals
117200010 |
1 g | Glass bottle |
chaque for $82.41
|
|
||||
AC117200050
|
Thermo Scientific Chemicals
117200050 |
5 g | Glass bottle |
chaque for $316.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
1499-10-1 | |
330.43 | |
FCNCGHJSNVOIKE-UHFFFAOYSA-N | |
15159 | |
9,10-diphenylanthracene |
C26H18 | |
MFCD00001253 | |
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard | |
CHEBI:51676 | |
C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 |
Spécifications
1499-10-1 | |
100.0 | |
Yellow to Beige | |
97.5% min. (HPLC) | |
C26H18 | |
1 g | |
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard | |
FCNCGHJSNVOIKE-UHFFFAOYSA-N | |
9,10-diphenylanthracene | |
15159 | |
330.42 | |
Fine Crystalline Powder |
97.5 | |
248°C to 252°C | |
Authentic | |
Glass bottle | |
MFCD00001253 | |
05,747 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,chloroform,benzene,carbon,disulfide,cyclohexane,acetonitrile,petroleum,ether | |
C1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12 | |
330.43 | |
CHEBI:51676 | |
98% | |
9, 10-Diphenylanthracene, 99% |
Sécurité et manipulation
missing translation for 'einecsNumber' : 216-105-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only