Biotix™ 96-Square V-Bottom Deep Well Microplates
Clear, stackable plates help save space in incubators and freezers.
$83.62 - $83.62
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
View More Specs
Products 1
Catalog Number Mfr. No. Sterility Price Quantity  
Catalog Number Mfr. No. Sterility Price Quantity  
12-111-077
View Documents
Biotix™
63300109
Sterile
Pack of 5 Each for $83.62
Case of 10 Pack for $836.20
Description
Description
  • Withstands centrifugation up to 6000 RCF
  • DNase, RNase and Endotoxin (pyrogen) free
Specifications
Specifications
No
Without Lid
Non Binding
Polypropylene
Non-treated
5
Standard
8 x 12
Clear
96
Square, Conical Bottom (V-well)
44 mm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'