missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.
Supplier: Eppendorf™ 0030129512
Description
- LoBind material guarantees maximum sample recovery for improved assay results.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
Specifications
Standard | |
8 x 12 | |
25 Plates (5 Bags x 5 Plates) | |
PCR Plate | |
Polypropylene, Polycarbonate | |
96 | |
PCR with low template concentration, Real-time PCR with low sample concentration, Low volume PCR/qPCR, NGS DNA library construction | |
PCR Clean (Non-sterile) | |
-80°C to +120°C |
Protect from direct sunlight and UV light; Store dry at room temperature | |
Autoclavable | |
Clear | |
Skirted | |
150 μL | |
Non-sterile | |
6000 x g | |
25 Pk. | |
Colorless |