missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ twin.tec™ PCR Plates LoBind™ - PCR Plates
Eppendorf twin.tec™ PCR plates LoBind, available in 96- and 384-well formats, maximize nucleic acid recovery and are certified as PCR clean for consistently reliable results.
$400.74 - $809.05
Specifications
Format | Standard |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene, Polycarbonate |
Sterility | Non-sterile |
Purity or Quality Grade | PCR Clean (Non-sterile) |
Catalog Number | Mfr. No. | No. of Wells | Volume (Metric) Well | Packaging | Color | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Wells | Volume (Metric) Well | Packaging | Color | Price | Quantity | ||
E0030129512
|
Eppendorf™
0030129512 |
96 | 150 μL | 25 Plates (5 Bags x 5 Plates) | Clear |
Pack of 25 Each for $432.39
|
|
||
14-380-052
|
Eppendorf™
0030531086 |
96 | 150 μL | 25 Plates (5 Bags x 5 Plates) | Clear |
Pack of 25 Each for $400.74
|
|
||
14-380-057
|
Eppendorf™
0030129882 |
96 | 150 μL | 25 Plates (5 Bags x 5 Plates) | Clear |
Pack of 25 Each for $400.74
|
|
||
14-380-054
|
Eppendorf™
0030531060 |
96 | 250 μL | 25 Plates (5 Bags x 5 Plates) | Clear |
Pack of 25 Each for $400.74
|
|
||
E0030129504
|
Eppendorf™
0030129504 |
96 | 250 μL | 25 Plates (5 Bags x 5 Plates) | Clear |
Pack of 25 Each for $432.39
|
|
||
14-380-053
|
Eppendorf™
0030531078 |
384 | 45 μL | 25 Plates (5 Bags x 5 Plates) | Clear |
Pack of 25 Each for $809.05
|
|
||
Description
- LoBind material guarantees maximum sample recovery for improved assay results.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
- Exceptionally solid and torque-resistant polycarbonate frame.
- OptiTrack™ matrix for faster sample identification and fewer pipetting errors.
- Raised well rims for effective sealing, also reduces risk of cross-contamination.
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean).
Specifications
Standard | |
Polypropylene, Polycarbonate | |
PCR Clean (Non-sterile) |
Autoclavable | |
Non-sterile |