Eppendorf™ twin.tec™ PCR Plates 96-well, 150 μL - PCR Plates
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. Raised well rims enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
Supplier:  Eppendorf™ 951020443
 View more versions of this product
Catalog No. E951020443
Explore available promotions
Description
Description
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
Specifications
Specifications
No Barcode
Standard
Autoclavable
Green
Skirted
150 μL
Non-sterile
2250 x g
25 Pk.
Human DNA, DNase, RNase-free
8 x 12
25 Plates (5 Bags x 5 Plates)
Standard PCR Plate
Polypropylene, Polycarbonate
96
Ideal for extreme applications such as multiple heat sealing.
PCR Clean (Non-sterile)
Colorless