missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Abietic acid, 85%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 173131000

 View more versions of this product

Catalog No. AC173131000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Abietic acid
514-10-3
100.0
85%
MFCD03423567
15, 7
RSWGJHLUYNHPMX-ONCXSQPRSA-N
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
10569
302.45
Technical
Authentic
Crystals and Chunks
85%
80.0
Brown-Yellow
C20H30O2
09, II, 424
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
302.46
CHEBI:28987
≥80%
100 g
Glass bottle

Chemical Identifiers

514-10-3
302.46
RSWGJHLUYNHPMX-ONCXSQPRSA-N
10569
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
C20H30O2
MFCD03423567
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
CHEBI:28987
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 208-178-3

RTECSNumber : TP8580000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only