missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Abiraterone Acetate 98.0+%, TCI America™

Supplier:  TCI America A28911G

 View more versions of this product

Catalog No. A28911G


Explore available promotions

Description

Description

Specifications

Specifications

Abiraterone Acetate
1 g
C26H33NO2
3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate
CC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CC=C4C5=CN=CC=C5)C)C
391.555
CHEBI:68639
≥98.0% (HPLC,T)
Powder/Crystals
154229-18-2
White/Yellow
MFCD00934213
UVIQSJCZCSLXRZ-UBUQANBQSA-N
[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
9821849
391.56
147°C

Chemical Identifiers

154229-18-2
391.555
UVIQSJCZCSLXRZ-UBUQANBQSA-N
9821849
[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
C26H33NO2
MFCD00934213
3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate
CHEBI:68639
CC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CC=C4C5=CN=CC=C5)C)C
Videos
Safety and Handling

Safety and Handling

RTECSNumber : BV7992100

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions