missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abiraterone Acetate 98.0+%, TCI America™
$503.46 - $1498.27
Chemical Identifiers
CAS | 154229-18-2 |
---|---|
Molecular Formula | C26H33NO2 |
Molecular Weight (g/mol) | 391.555 |
MDL Number | MFCD00934213 |
InChI Key | UVIQSJCZCSLXRZ-UBUQANBQSA-N |
Synonym | 3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate |
PubChem CID | 9821849 |
ChEBI | CHEBI:68639 |
IUPAC Name | [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
SMILES | CC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CC=C4C5=CN=CC=C5)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
A2891200MG
|
TCI America
A2891200MG |
200 mg |
Each for $503.46
|
|
||||||
A28911G
|
TCI America
A28911G |
1 g |
Each for $1,498.27
|
|
||||||
Chemical Identifiers
154229-18-2 | |
391.555 | |
UVIQSJCZCSLXRZ-UBUQANBQSA-N | |
9821849 | |
[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
C26H33NO2 | |
MFCD00934213 | |
3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate | |
CHEBI:68639 | |
CC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CC=C4C5=CN=CC=C5)C)C |
Specifications
154229-18-2 | |
White/Yellow | |
MFCD00934213 | |
UVIQSJCZCSLXRZ-UBUQANBQSA-N | |
[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate | |
9821849 | |
391.56 | |
147°C | |
Abiraterone Acetate |
200 mg | |
C26H33NO2 | |
3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate | |
CC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CC=C4C5=CN=CC=C5)C)C | |
391.555 | |
CHEBI:68639 | |
≥98.0% (HPLC,T) | |
Powder/Crystals |
Safety and Handling
RTECSNumber : BV7992100
TSCA : No
Recommended Storage : Freezer