missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Accessories for Orion™ pH Electrodes

Store and protect your pH electrodes with these accessories.

Supplier:  Thermo Scientific™ 910003WA

 View more versions of this product

Catalog No. 13-641-42


Explore available promotions

Includes: 3 Electrode Storage Bottles

Description

Description

Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.

Electrode Storage Bottles

  • Can be used with any 6, 8 or 12mm pH electrodes
Specifications

Specifications

12mm electrode storage bottle
12 mm Electrodes, Glass
Electrode Storage Bottle
Videos
SDS
Documents

Documents

Promotions

Promotions