Fisherbrand™ accumet™ Liquid-Filled Polymer Body Combination Electrodes: Mercury-Free
Ideal for measurements in small vessels or with small samples
Supplier:  Fisherbrand™ 13620220
 View more versions of this product
Catalog No. 13-620-220
Explore available promotions
Description
Description
Ideal for measurements in small vessels or with small samples
Standard pH Combination Electrodes
  • Rugged polypropylene body—ideal for handheld use with aqueous samples
  • Endcap seal keeps membrane moist and ready
  • 0 to 14pH range
  • All have porous ceramic junction
  • Available with single-junction Ag/AgCl reference or with double-junction Ag/AgCl reference
Specifications
Specifications
U.S. Standard/Pin
Polypropylene
-5°C to +100°C
Saturated KCl with AgCl. KCl
Ag/AgCl
-5°C to +100°C
106 mm
10 mm
Porous Ceramic Double
0 to 14
pH Combination Electrode
For measurements in small vessels or with small samples
23°F to 212°F
4.1 in.
0.4 in.