missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acebutolol hydrochloride, Thermo Scientific Chemicals

Cardioselective beta-adrenergic blocker

Supplier:  Thermo Scientific Chemicals J6379003

 View more versions of this product

Catalog No. AAJ6379003


Explore available promotions

Description

Description

Cardioselective beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acebutolol hydrochloride
141°C to 143°C
C18H29ClN2O4
1 g
acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn
[H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O
372.89
CHEBI:2380
Powder
34381-68-5
White
MFCD00078860
14,19
KTUFKADDDORSSI-UHFFFAOYNA-N
hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride
441307
372.89

Chemical Identifiers

34381-68-5
372.89
KTUFKADDDORSSI-UHFFFAOYNA-N
441307
hydrogen N-(3-acetyl-4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)butanamide chloride
C18H29ClN2O4
MFCD00078860
acebutolol hydrochloride, acebutolol hcl, n-3-acetyl-4-2-hydroxy-3-isopropylamino propoxy phenyl butyramide hydrochloride, diasectral, wesfalin, 3'-acetyl-4'-2-hydroxy-3-isopropylamino propoxy butyranilide hydrochloride, dl-1-2-acetyl-4-butyramidophenoxy-2-hydroxy-3-isopropylaminopropane hydrochloride, ccris 1102, sectral tn
CHEBI:2380
[H+].[Cl-].CCCC(=O)NC1=CC=C(OCC(O)CNC(C)C)C(=C1)C(C)=O
Safety and Handling

Safety and Handling

EINECSNumber : 251-980-3

RTECSNumber : ES5235000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only